Skip to Content
Merck

Skip To

PPB022

Tris buffer

Pre-set, pH 7.0, pHast Pack, powder

Synonym(s):

0.1M Tris

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 pkg
Please contact Customer Service for Availability
2 640,00 CZK

About This Item

NACRES:
NA.25
UNSPSC Code:
12161700

2 640,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

pHast Pack

form

powder

storage condition

(Tightly closed. Dry)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

7.0

solubility

soluble, clear, colorless

cation traces

Fe: ≤10 ppm

foreign activity

DNase, none detected, Endonuclease, none detected, Exonuclease, none detected, NICKase, none detected, Protease, none detected, RNase, none detected

storage temp.

room temp

General description

Tris buffer, pH 7.0 is a general buffer that is isotonic and non-toxic in nature due to the presence of tris base. It is suitable for most biochemical and molecular biology applications. Tris has a buffering capacity range of pH 7.0 - 9.0, with a pKa of 8.1 at 25°C.

Application

Tris buffer is used in:
  • Western blot
  • Protein gel electrophoresis
  • cell culture (sterile filter buffer)
  • As a buffer component

Features and Benefits

  • Fast, ready-to-use buffer
  • No need to measure and adjust the pH
  • Saves time and minimizes effort in the lab
  • Biological tests: free of DNase, RNase, Protease, and Nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Preparation Note

Contents of one pouch, when dissolved in 500 mL of ultrapure water, will yield a 0.1M Tris solution, pH 7.0 at 25 °C. Contents tested to be DNAse, RNAse, Protease, and Nickase free. This product is suitable for use in cell culture and electrophoresis applications.
Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PPB023PPB015PPB011
description

Pre-set, pH 7.0, pHast Pack, powder

description

Pre-set, pH 8.1, pHast Pack, powder

description

pH 8.3, pHast Pack, powder

description

pH 7.6, pHast Pack, powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

product line

pHast Pack

product line

pHast Pack

product line

pHast Pack

product line

pHast Pack

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage condition

(Tightly closed. Dry)

storage condition

(Tightly closed. Dry)

storage condition

(Tightly closed. Dry)

storage condition

(Tightly closed. Dry)

pH

7.0

pH

8.1

pH

8.3

pH

7.6


Still not finding the right product?

Explore all of our products under Tris buffer


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters