Skip to Content
Merck
All Photos(1)

Key Documents

PHR2867

Supelco

Fluorouracil Related Compound E

certified reference material, pharmaceutical secondary standard

Synonym(s):

5-Chlorouracil, Fluorouracil Impurity E (PhEur), 5-Chloro-2,4-dihydroxypyrimidine, 5-Chloropyrimidine-2,4(1H,3H)-dione

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 14,500.00

CZK 14,500.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
CZK 14,500.00

About This Item

Empirical Formula (Hill Notation):
C4H3ClN2O2
CAS Number:
Molecular Weight:
146.53
Beilstein:
127173
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 14,500.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1279054

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

fluorouracil

form

solid

packaging

pkg of 50 mg

mp

>300 °C (lit.)

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

SMILES string

ClC1=CNC(=O)NC1=O

InChI

1S/C4H3ClN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

ZFTBZKVVGZNMJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service