form
solid
shelf life
expiry date on the label
mol wt
Mr 2500-5000 Da
technique(s)
MALDI-MS: suitable
storage temp.
−20°C
General description
SpheriCal™ is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal™ helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibrations, SpheriCal™ offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.
Mass ranges available:
i.SpheriCal™ Aqua peptide low (m/z 500 - 1,000)
ii.SpheriCal™ Aqua peptide medium (m/z 1,000 - 2,500)
iii.SpheriCal™ Aqua peptide high (m/z 2,500 - 5,200)
iv.SpheriCal™ Aqua protein low (m/z 5,200 - 11,000)
SpheriCal™ Aqua Neat: ideal for water-based systems.
SpheriCal™ Aqua is ideal for calibration with aqueous systems commonly used in life sciences, e.g., matrices DHB, HCCA and SA or solvents such as water and acetonitrile, although ionization is preferred with alkali cations. Each vial of SpheriCal™ Neat contains sufficient calibration standard for over 100 calibration spots.
Find more information here SpheriCal™
Mass ranges available:
i.SpheriCal™ Aqua peptide low (m/z 500 - 1,000)
ii.SpheriCal™ Aqua peptide medium (m/z 1,000 - 2,500)
iii.SpheriCal™ Aqua peptide high (m/z 2,500 - 5,200)
iv.SpheriCal™ Aqua protein low (m/z 5,200 - 11,000)
SpheriCal™ Aqua Neat: ideal for water-based systems.
SpheriCal™ Aqua is ideal for calibration with aqueous systems commonly used in life sciences, e.g., matrices DHB, HCCA and SA or solvents such as water and acetonitrile, although ionization is preferred with alkali cations. Each vial of SpheriCal™ Neat contains sufficient calibration standard for over 100 calibration spots.
Find more information here SpheriCal™
dendritic calibrants PFQ3A : PFQ3B : PFQ3C : PFQ3D = 1:1:1:1
Application
For mass calibration in mass spectrometry
Legal Information
SpheriCal is a trademark of Polymer Factory Sweden AB
Still not finding the right product?
Explore all of our products under SpheriCal™ Aqua
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.