Skip to Content
Merck
All Photos(1)

Key Documents

P9750

Sigma-Aldrich

Phenolphthalein

pH indicator

Synonym(s):

3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O4
CAS Number:
Molecular Weight:
318.32
Colour Index Number:
764
Beilstein:
284423
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

technique(s)

titration: suitable

mp

261-263 °C (lit.)

solubility

acetone: soluble

SMILES string

Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H

InChI key

KJFMBFZCATUALV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 105945-Sigma-Aldrich | Phenolphthalein ACS reagent

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Bormashenko et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(4), 654-656 (2009-01-30)
The mechanisms of floating and sliding of liquid marbles are studied. Liquid marbles containing CaCl(2) and marbles containing NaOH water solutions float on water containing Na(2)CO(3) and an alcoholic solution of phenolphthalein with no chemical reaction. Sliding of liquid marbles
Tamer Uyar et al.
ACS nano, 4(9), 5121-5130 (2010-08-20)
Electrospinning has been used to successfully create polystyrene (PS) nanofibers containing either of three different types of cyclodextrin (CD); α-CD, β-CD, and γ-CD. These three CDs are chosen because they have different sized cavities that potentially allow for selective inclusion
Ping Chen et al.
ACS nano, 5(7), 5928-5935 (2011-06-15)
In this paper, we report the fabrication of carbonaceous nanofiber (CNF) membranes functionalized by beta-cyclodextrins (CNF-β-CD membrane) and their application for molecular filtration. The chemically synthesized carbonaceous nanofibers were first functionalized by β-CD, and the free-standing CNF membrane can be
Pieter A Cohen et al.
Journal of general internal medicine, 27(1), 51-56 (2011-08-17)
Pai You Guo is a weight loss supplement manufactured in China and adulterated with the banned pharmaceutical products sibutramine and phenolphthalein. The US Food and Drug Administration (FDA) announced a voluntary recall of Pai You Guo in 2009, yet clinicians
Nantao Hu et al.
Journal of nanoscience and nanotechnology, 12(1), 99-104 (2012-04-25)
Recently, much work has focused on the exfoliation of graphene through a combination of oxidation and sonication procedures, followed by reduction through chemical methods. We demonstrated that the individual graphene oxide sheets can be readily reduced by using phenolphthalin as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service