Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 g | Please contact Customer Service for Availability | CZK 5,440.00 |
About This Item
CZK 5,440.00
agency
according to ISO 6579:2014
Quality Segment
sterility
non-sterile
product line
BioChemika
form
powder
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbe id | utilization test: suitable, microbiological culture: suitable
final pH
7.4±0.2 (25 °C)
application(s)
clinical testing
food and beverages, microbiology
suitability
nonselective and differential for bacteria (General Media), nonselective and differential for enterobacteriaceae
Application
Preparation Note
Other Notes
Proteose peptone, 10.00
Beef extract, 1.00
Sodium chloride, 5.00
Phenol red, 0.018
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| agency according to ISO 6579:2014 | agency - | agency - | agency according to ISO 11290-1:2017, according to ISO 11290-2:2017 |
| suitability nonselective and differential for bacteria (General Media), nonselective and differential for enterobacteriaceae | suitability nonselective and differential for Citrobacter spp., nonselective and differential for Enterobacter spp., nonselective and differential for Escherichia coli, nonselective and differential for Klebsiella spp., nonselective and differential for Proteus spp., nonselective and differential for Salmonella spp., nonselective and differential for Serratia spp., nonselective and differential for Shigella spp., nonselective and differential for coliforms, nonselective and differential for enterobacteriaceae | suitability bacteria, nonselective and differential for Enterobacter spp., nonselective and differential for Escherichia coli, nonselective and differential for Klebsiella spp., nonselective and differential for Salmonella spp., nonselective and differential for Shigella spp., nonselective and differential for coliforms | suitability Enterococcus spp., Haemophilus spp., Pneumococcus spp., nonselective and differential for Campylobacter spp., nonselective and differential for Neisseria spp., nonselective and differential for Staphylococcus spp., nonselective and differential for Streptococcus spp., nonselective and differential for bacteria (General Media), nonselective and differential for molds (General Media), nonselective and differential for yeasts (General Media) |
| technique(s) microbe id | utilization test: suitable, microbiological culture: suitable | technique(s) microbe id | specific enzyme detection: suitable, microbiological culture: suitable | technique(s) microbe id | utilization test: suitable, microbiological culture: suitable | technique(s) microbe id | specific enzyme detection: suitable, microbiological culture: suitable |
| form powder | form powder | form powder | form powder |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| application(s) clinical testing | application(s) clinical testing | application(s) food and beverages, microbiology | application(s) clinical testing |
Still not finding the right product?
Explore all of our products under Phenol red broth base
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



