Skip to Content
Merck
All Photos(1)

Key Documents

NIST887

Cemented carbide (W-83, Co-10)

NIST SRM 887

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 16,800.00

CZK 16,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 16,800.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 16,800.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is a sintered tungsten carbide base material in the form of a fine powder (150μm) intended for use in checking chemical and instrumental methods of analysis. For more information, please refer to the SDS and the COA.

SRM 887_cert

SRM 887_SDS

Other Notes

Certified for the analytes listed below.
Cobalt (Co)

Reference/Informational value is provided for the following analytes.
Molybdenum (Mo), Nickel (Ni), Iron (Fe), Titanium (Ti), Tantalum (Ta), Niobium (Nb), Carbon (C)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service