Skip to Content
Merck
All Photos(1)

Key Documents

NIST8664

Ginseng-Containing Solid Oral Dosage Form

NIST®SRM®

Synonym(s):

Herbal supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 13 g

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of RM 8664 includes five packets, each measuring ~ 2.6 grams of ground ginseng-containing solid oral dosage form.
For more information, please refer to the SDS and COA.
SRM 8664_cert
SRM 8664_SDS

Application
This Standard Reference Material (RM) is intended for use in evaluating the analytical methods employed to determine ginsenosides in solid oral dosage forms of ground ginseng and similar matrices.

Features and Benefits

  • Available with non-certified mass fractionvalues for ginsenosides, provided on a dry-mass basis and values aremetrologically traceable to the materials and methods employed in theirdetermination.
  • Period of validity, source, instructions for storage, handling, and uses areprovided on the NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Ginsenoside Rb1 (C54H92O23), Ginsenoside Rb2 (C53H90O22), Ginsenoside Rc (C53H90O22), Ginsenoside Rd (C53H90O22), Ginsenoside Re (C53H90O22), Ginsenoside Rf (C53H90O22), Ginsenoside Rg1 (C42H72O14)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service