Skip to Content
Merck
All Photos(1)

Documents

NIST656

Silicon nitride powders

NIST® SRM® 656, for quantitative analysis by powder diffraction

Synonym(s):

Silicon nitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

powder

packaging

pkg of 2 x 10 g

manufacturer/tradename

NIST®

density

3.44 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service