Skip to Content
Merck
All Photos(1)

Key Documents

NIST58A

Ferrosilicon

NIST® SRM® 58a, 73% Si - regular grade

Sign Into View Organizational & Contract Pricing

Select a Size

75 G
CZK 22,000.00

CZK 22,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
75 G
CZK 22,000.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 22,000.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 75 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in validation of chemical and instrumental methods of analysis. A unit of SRM 58a consists of a bottle containing approximately 75 g of fine powder. For more information, please refer to the SDS and COA.

SRM 58A_cert

SRM 58A_SDS

Other Notes

Certified for the analytes listed below.
Copper (Cu), Chromium (Cr), Iron (Fe), Manganese (Mn), Nickel (Ni), Silicon (Si)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Carbon (C), Calcium (Ca), Phosphorus (P), Titanium (Ti), Arsenic (As), Boron (B), Cobalt (Co), Molybdinum (Mo), Oxygen (O), Sulfur (S), Tin (Sn), Vanadium (V), Zirconium (Zr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service