Skip to Content
Merck
All Photos(1)

Documents

NIST3672

Organic contaminants in smokers′ urine (frozen)

NIST® SRM® 3672

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

application(s)

clinical research

format

matrix material

storage temp.

−70°C

General description

Each unit of SRM 3672 consists of five vials each containing 10 mL of frozen urine.

SRM 3672 _SDS

SRM 3672 _CoA

Application

Organic contaminants in smokers′ urine standard reference material (SRM) is intended for use in evaluating analytical methods used to determine selected hydroxylated polycyclic aromatic hydrocarbons (hydroxylated PAHs) and phthalate, phenol, and volatile organic compound (VOC) metabolites in urine.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of the hydroxylated PAHs.
  • Certified values are metrologically traceable to the International System of Units (SI) and SI-derived units of mass fraction.   
  • The non-certified values of phthalate, phenol, VOC metabolites, and organic analytes are supplied in the NIST certificate. 
  • The NIST certificate is provided with storage, use and safety instructions.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service