Skip to Content
Merck
All Photos(1)

Documents

NIST278

Obsidian rock

NIST® SRM® 278

Synonym(s):

Obsidian

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 35 g

manufacturer/tradename

NIST®

format

matrix material

General description

It is a finely powdered obstian rock, derived from Clear Lake, Newberry Crater, Oregon.

SRM 278_cert

SRM 278_SDS

Application

Obsidian rock standard reference material (SRM) is intended for assessing the precision of both analytical methods and instruments employed in analyzing geological materials.

Features and Benefits

  • Available with a certificate documenting NIST-certified values of constituents in the obsidian rock, these values were determined through either a definitive method, reference method, or two or more analytical methods.
  • Information values of the constituents are determined by single or non-reference method.

Other Notes

Certified for the analytes listed below.
Aluminium dioxide (Al2O3), Calcium oxide (CaO), Iron(II) oxide (FeO), Iron(III) oxide (Fe2O3), Potassium oxide (K2O), Manganese oxide (MnO), Sodium oxide (Na2O), Phosphorus pentoxide (P2O5), Silicon dioxide (SiO2), Titanium dioxide (TiO2), Copper (Cu), Nickel (Ni), Rubidium (Rb), Strontium (Sr), Thorium (Th), Thallium (Tl), Uranium (U), Lead (Pb)

Reference/Informational value is provided for the following analytes.
Carbon (C), Carbon dioxide (CO2), Fluorine (F), Magnesium oxide (MgO), Barium (Ba), Boron (B), Cerium (Ce), Cobalt (Co), Chromium (Cr), Caesium (Cs), Europium (Eu), Gadolinium (Gd), Hofnium (Hf), Lutetium (Lu), Antimony (Sb), Scandium (Sc), Samarium (Sm), Tantalum (Ta), Terbium (Tb), Ytterbium (Yb), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reference samples for use in analytical geochemistry: their availability, preparation, and appropriate use.
Kane J S.
Journal of Geochemical Exploration, 44(1-3), 37-63 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service