Skip to Content
Merck
All Photos(1)

Documents

NIST2693

Bituminous coal (sulfur, mercury, and chlorine)

NIST® SRM® 2693

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in the evaluation of techniques employed in the determination of sulfur, mercury, chlorine, and ash content in coal and materials of a similar matrix. A unit of SRM 2693 consists of 50 g of bituminous coal ground to pass a 250 μm (60 mesh) sieve, homogenized, and packaged in an amber glass bottle and then sealed in an aluminized bag. For more information, please refer to the SDS and COA.

SRM 2693_cert

SRM 2693_SDS

Other Notes

Certified for the analytes listed below.
Sulfur (S), Mercury (Hg), Chlorine (Cl)

Reference/Informational value is provided for the following analytes.
Ash content

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service