Skip to Content
Merck
All Photos(1)

Key Documents

NIST2572

Lead paint film for portable x-ray fluorescence analyzers

NIST® SRM® 2572, nominal 1.6 mg/cm2 (color code:  orange)

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
CZK 16,000.00

CZK 16,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
CZK 16,000.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 16,000.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 1 +blank

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

coatings

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for validation of results from portable, hand-held, X-ray fluorescence analyzers, when testing for lead in paint coatings on interior and exterior building surfaces. A unit of SRM 2572 consists of a white polyester sheet, approximately 7.6 cm wide, 10.2 cm long, and 0.2 mm thick, coated with a single, orange-colored paint layer, approximately 0.04 mm thick. Included is one unit of SRM 2570, which is coated with a lead-free, lacquer layer of the same thickness as a lead paint layer and is intended as a blank. All sheets are over-coated with a clear, thin, plastic laminate to protect the surface from abrasion. For more information, please refer to the COA and SDS.

SRM 2572_cert

SRM 2572_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service