Skip to Content
Merck

Skip To

NIST1908

Vickers microhardness of nickel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
41 000,00 CZK

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

41 000,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

packaging

pkg of each

manufacturer/tradename

NIST®

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nickel (Ni)

See certificate for values and more details at nist.gov/SRM.

Legal Information

SRM is a registered trademark of National Institute of Standards and Technology
NIST is a registered trademark of National Institute of Standards and Technology

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NIST1909NIST2829NIST2798A
description

-

description

-

description

NIST® SRM® 2829

description

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

packaging

pkg of each

packaging

pkg of each

packaging

pkg of each

packaging

pkg of each

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Vickers microhardness of nickel


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters