Skip to Content
Merck
All Photos(1)

Key Documents

NIST1255B

Aluminum alloy 356

NIST® SRM® 1255b

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
CZK 40,000.00

CZK 40,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
CZK 40,000.00

About This Item

UNSPSC Code:
12352311
NACRES:
NA.24

CZK 40,000.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1255b contains a disk with a diameter of approximately 6.3 cm and a thickness of 1.9 cm.

SRM 1255B_cert

SRM 1255B_SDS

Application

The SRM is primarily intended for evaluating the method of analyzing elements in aluminum alloys.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified massfraction values, expanded uncertainties, and coverage factor (k) for elements in SRM 1255b.
  • The certified values are metrologically traceable to the SI derived unit of mass fraction expressed as percent.
  • The reference value of chromium is reported as mass fraction value.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Silicon (Si), Iron (Fe), Copper (Cu), Manganese (Mn), Magnesium (Mg), Nickel (Ni), Zinc (Zn), Titanium A (Ti), Titanium B (Ti), Vanadium A (V), Vanadium B (V), Strontium A (Sr), Strontium B (Sr), Gallium (Ga), Tin (Sn), Lead (Pb)

Reference/Informational value is provided for the following analytes.
Chromium (Cr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service