Skip to Content
Merck
All Photos(1)

Documents

NIST1132

Lead-base bearing metal (84Pb - 10Sb - 6Sn)

NIST® SRM® 1132

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

technique(s)

X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable

format

matrix material

General description

This Standard Reference Material is in disk form, 32 mm (1 1/4 in.) in diameter and 19 mm (3/4 in.) thickness. It is intended primariliy for use in optical emission and x-ray spectrometric methods of analysis. For more information, please refer to the COA ad SDS.

SRM 1132_cert

SRM 1132_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Antimony (Sb), Tin (Sn), Copper (Cu), Bismuth (Bi), Arsenic (As), Nickle (Ni), Iron (Fe)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1 Oral - STOT RE 2 - STOT RE 2 Inhalation

Target Organs

Central nervous system,Blood,Immune system,Kidney, Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service