Skip to Content
Merck
All Photos(1)

Key Documents

NIST1010A

Microcopy resolution test charts

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
CZK 16,800.00

CZK 16,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 EA
CZK 16,800.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 16,800.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of set (5)

manufacturer/tradename

NIST®

application(s)

general analytical

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service