Skip to Content
Merck
All Photos(1)

Key Documents

F171703

Sigma-Aldrich

Diamond 1000 μL tip pack

for use with BioControl® GDS, suitable for PCR, tip volume 1000 μL

Synonym(s):

Diamond 1000 μL tip pack, Sterile Pipette Tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 EACH
CZK 2,350.00

CZK 2,350.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
960 EACH
CZK 2,350.00

About This Item

UNSPSC Code:
41106302
NACRES:
NB.02

CZK 2,350.00


Please contact Customer Service for Availability

Request more information

sterility

sterile

packaging

box of 10 × 96 units

technique(s)

PCR: suitable

tip volume

1000 μL

compatibility

for use with BioControl® GDS

storage temp.

room temp

General description

Disposable tips are used to transfer PCR reagents. They can be used as consumable accessories during GDS PCR sample preparation.

Application

The Assurance® GDS Diamond Tips are used to transfer liquids during Assurance® GDS sample preparation.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service