Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
format
matrix material
storage temp.
2-8°C
General description
Certified for gross calorific value (GCV), net calorific value (NCV), C, N, S, Ca, Na, Se, Zn.
Analysis Note
For more information please see:
ERMEF413
ERMEF413
Legal Information
ERM is a registered trademark of European Commission
1 of 1
This Item | |||
|---|---|---|---|
| format matrix material | format matrix material | format matrix material | format matrix material |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| agency ERM® | agency ERM® | agency ERM® | agency ERM® |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
