Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
application(s)
petroleum
format
matrix material
Analysis Note
please visit the JRC webpage
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description ERM®, certified reference material | description ERM®, certified reference material | description BCR®, certified reference material | description BCR®, certified reference material |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| format matrix material | format matrix material | format matrix material | format matrix material |
| application(s) petroleum | application(s) petroleum | application(s) environmental | application(s) environmental |
| agency ERM® | agency ERM® | agency BCR® | agency BCR® |
Still not finding the right product?
Explore all of our products under Diesel (B7)
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
134.6 °F
flash_point_c
57 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




