Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
agency
ERM®, certified by the BSC
manufacturer/tradename
JRC
application(s)
clinical testing
format
matrix material
storage temp.
−20°C
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Other Notes
Sales restrictions may apply
Legal Information
ERM is a registered trademark of European Commission
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.