Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | 4 400,00 CZK |
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
application(s)
general analytical
format
matrix material
General description
Certified for the mass fractions of As, Cd, Cu, Hg, Pb, Se and Zn
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Other Notes
Matrix Group: Fruits and Vegetables
Legal Information
ERM is a registered trademark of European Commission
1 of 1
This Item | |||
|---|---|---|---|
| description ERM®, certified reference material | description ERM®, certified reference material | description ERM®, certified reference material | description ERM®, certified reference material |
| application(s) general analytical | application(s) - | application(s) food and beverages | application(s) - |
| format matrix material | format matrix material | format matrix material | format matrix material |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| agency ERM® | agency ERM® | agency ERM® | agency ERM® |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
