Skip to Content
Merck
All Photos(1)

Key Documents

E3258

Supelco

Ethinamate

analytical standard

Synonym(s):

1-Ethynylcyclohexanol carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

NC(=O)OC1(CCCCC1)C#C

InChI

1S/C9H13NO2/c1-2-9(12-8(10)11)6-4-3-5-7-9/h1H,3-7H2,(H2,10,11)

InChI key

GXRZIMHKGDIBEW-UHFFFAOYSA-N

Gene Information

human ... CA1(759)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Urethane derivative with rapid onset and short duration of action.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drugs in dentistry. Sedatives.
S G Ciancio
Dental management, 25(4), 50-50 (1985-04-01)
The metabolism of drugs and other foreign compounds in suspensions of isolated rat hepatocytes.
R E McMahon
Annals of the New York Academy of Sciences, 349, 46-56 (1980-01-01)
A simultaneous plasma level and EEG assessment of an oral hypnotic (ethinamate).
C F Speirs et al.
European journal of pharmacology, 20(1), 127-129 (1972-10-01)
M I Walash et al.
Journal - Association of Official Analytical Chemists, 68(6), 1209-1212 (1985-11-01)
Titrimetric and spectrophotometric titration methods are described for the quantitative determination of acetylenic hypnotics ethchlorvynol, ethinamate, and meparfynol carbamate as pure substances and in dosage forms. The methods involve the use of different brominating agents. A known excess of the
A el-Brashy et al.
Pharmaceutisch weekblad. Scientific edition, 10(2), 90-92 (1988-04-22)
A titrimetric method is described for the determination of three acetylenic hypnotics, namely ethchlorvynol, ethinamate, and methylpentynol carbamate, in bulk and in dosage forms. The method involves the use of either 1,3-dibromo-5,5-dimethylhydantoin (DBH), N-bromosuccinimide (NBS) or N-bromophthalimide (NBP) as titrants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service