Skip to Content
Merck
All Photos(1)

Key Documents

DLW354442

Corning® BioCoat Insert 6w 0.4um Col I 24cas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
Pricing and availability is not currently available.

shipped in

wet ice

storage temp.

2-8°C

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service