Skip to Content
Merck
All Photos(1)

Key Documents

D1250000

Diethanolamine fusidate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H59NO8
CAS Number:
Molecular Weight:
621.84

application(s)

pharmaceutical (small molecule)

InChI

1S/C31H48O6.C4H11NO2/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;6-3-1-5-2-4-7/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);5-7H,1-4H2/b26-20-;/t18-,21-,22-,23+,24+,25-,27?,29-,30-,31-;/m0./s1

InChI key

OQZZCXRLEFBPLZ-SQCIRFEJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Engelen et al.
The Veterinary record, 147(20), 567-569 (2000-12-05)
Eighty-nine cats and 38 dogs naturally infested with the ear mite Otodectes cynotis were randomly allocated into two treatment groups. One group was treated with a product containing miconazole nitrate, polymyxin B sulphate and prednisolone acetate, the other with a
R Bayston et al.
The Journal of bone and joint surgery. British volume, 64(4), 460-464 (1982-01-01)
The release of gentamicin sulphate, sodium fusidate and diethanolamine fusidate from Palacos and CMW cements was studied using elution and serial plate transfer tests. Further tests were made to assay the drug remaining in the cement after antibacterial activity could
Diethanolamine fusidate has no in-vitro activity against the human immunodeficiency virus.
C Birch et al.
The Medical journal of Australia, 149(4), 224-225 (1988-08-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service