Skip to Content
Merck

Skip To

BP797

Fluoxetine hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mg
Please contact Customer Service for Availability
CZK 6,310.00

About This Item

Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
260-101-2
MDL number:

CZK 6,310.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

fluoxetine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CNCCC(C1=CC=CC=C1)OC2=CC=C(C(F)(F)F)C=C2.[H]Cl

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H

InChI key

GIYXAJPCNFJEHY-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluoxetine hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Fluoxetine Oral Solution
  • Fluoxetine Capsules
  • Biochem/physiol Actions

    Selective serotonin reuptake inhibitor; antidepressant.

    Packaging

    Unit quantity: 250 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Legal Information

    Sold under license from Eli Lilly and Company.
    Prozac is a registered trademark of Eli Lilly and Co.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    340121279804343290
    grade

    pharmaceutical primary standard

    grade

    analytical standard

    grade

    pharmaceutical primary standard

    grade

    -

    manufacturer/tradename

    BP

    manufacturer/tradename

    -

    manufacturer/tradename

    USP

    manufacturer/tradename

    Calbiochem®

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    forensics and toxicology
    pharmaceutical (small molecule)

    application(s)

    pharmaceutical (small molecule)

    application(s)

    -

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    -

    shelf life

    -

    form

    solid

    form

    -

    form

    -

    form

    solid

    format

    neat

    format

    neat

    format

    neat

    format

    -


    Still not finding the right product?

    Explore all of our products under Fluoxetine hydrochloride


    signalword

    Danger

    Hazard Classifications

    Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

    target_organs

    Central nervous system

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters