Skip to Content
Merck

Skip To

BP650

Terfenadine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

α-[4-(1,1-Dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinebutanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Please contact Customer Service for Availability
CZK 6,040.00

About This Item

Empirical Formula (Hill Notation):
C32H41NO2
CAS Number:
Molecular Weight:
471.67
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

CZK 6,040.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

terfenadine

form

crystalline powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C32H41NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3

InChI key

GUGOEEXESWIERI-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Terfenadine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Biochem/physiol Actions

Non-sedating second generation H1 histamine receptor antagonist.
Non-sedating second generation H1 histamine receptor antagonist. Mainly metabolized by Cyp3A4, 5, 7. Inhibits CYP2C8.

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T0071000T9652T0080000
manufacturer/tradename

BP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

form

crystalline powder

form

-

form

-

form

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Terfenadine


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters