Skip to Content
Merck

BCR166

Latex spheres

BCR®, certified reference material, 4.8 μm particle size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

BCR®

description

parameter of the calibration line 4.821 0.019

form

particles

manufacturer/tradename

JRC

particle size

4.8 μm

application(s)

general analytical

format

neat

General description

Nominal 4.8 μm latex (0.2 % solids)

Packaging

Vial containing 2 mL of an aqueous suspension of latex spheres.

Analysis Note

For more information please see:
BCR166

Legal Information

BCR is a registered trademark of European Commission


Still not finding the right product?

Explore all of our products under Latex spheres


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library