Skip to Content
Merck
All Photos(1)

Key Documents

A9906

Supelco

Amino acid standards, physiological

analytical standard, acidics, neutrals, and basics

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
85151701
eCl@ss:
32160406
NACRES:
NA.24
To inquire about this product A9906, please contact your local Merck office or dealer. Contact Technical Service

grade

analytical standard

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

The following components are included in the amino acid standard:
  • L-Alanine
  • Ammonium chloride
  • L-Arginine
  • L-Aspartic acid
  • L-Cystine
  • L-Glutamic acid
  • Glycine
  • L-Histidine
  • L-Isoleucine
  • L-Leucine
  • L-Lysine
  • L-Methionine
  • L-Phenylalanine
  • L-Proline
  • L-Serine
  • L-Threonine
  • L-Tyrosine
  • L-Valine
  • β-Alanine
  • L-α-Amino-n-butyric acid
  • γ-Amino-n-butyric acid
  • DL-β-Aminoisobutryic acid
  • L-Anserine
  • L-Carnosine
  • L-Citrulline
  • L-Creatinine
  • Cystathionine
  • Ethanolamine
  • L-Homocystine
  • δ-Hydroxylysine
  • Hydroxy-L-proline
  • 1-Methyl-L-histidine
  • 3-Methyl-L-histidine
  • L-Ornithine
  • Sarcosine
  • Taurine
  • L-Tryptophan
  • Urea

Other Notes

Amino acids and related compounds are 0.5 μmole/mL in 0.2 N lithium citrate, pH 2.2 containing 0.1% phenol and 2% thiodiglycol.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petronia Carillo et al.
Food chemistry, 292, 166-175 (2019-05-06)
Compositional characterization was performed on seven 'Pomodorino del Piennolo del Vesuvio' (PPV) tomato landraces, a signature product of Campania (Italy) threatened by genetic erosion. Characterization encompassed determinations of macro-minerals, soluble carbohydrates, starch, acidity, lycopene, polyphenols, anthocyanins, protein and free amino
Patricia Mitry et al.
European journal of clinical nutrition, 73(5), 692-702 (2018-07-19)
Dietary intake of red and processed meat has been associated with disease risk. Since dietary intake assessment methods are prone to measurement errors, identifying biomarkers of meat intake in bio-samples could provide more valid intake estimates. We examined associations of
Eli K Moore et al.
Frontiers in microbiology, 6, 637-637 (2015-07-16)
Microbial decomposition of organic matter is an essential process in the global carbon cycle. The soil bacteria Pseudopedobacter saltans and Flavobacterium johnsoniae are both able to degrade complex organic molecules, but it is not fully known how their membrane structures
Huibin Qin et al.
Frontiers in microbiology, 7, 1311-1311 (2016-09-10)
Suanzhou as a traditional Chinese gruel is fermented from proso millet and millet. The biochemical analysis showed Suanzhou had relatively high concentrations of lactic acid, acetic acid, and free amino acids. The metagenomics of Suanzhou were studied, with the analysis
Diogo Antonio Alves de Vasconcelos et al.
The Journal of nutritional biochemistry, 70, 202-214 (2019-06-25)
L-Glutamine (L-Gln) supplementation has been pointed out as an anticatabolic intervention, but its effects on protein synthesis and degradation signaling in skeketal muscle are still poorly known. The effects of L-Gln pretreatment (1 g kg-1 day-1 body weight for 10

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service