Skip to Content
Merck

96848

Methyl crotonate

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH=CHCOOCH3
CAS Number:
Molecular Weight:
100.12
EC Number:
210-793-7
UNSPSC Code:
85151701
PubChem Substance ID:
Beilstein/REAXYS Number:
1720292
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤1.5% water

refractive index

n20/D 1.423 (lit.), n20/D 1.424-1.426

bp

118-120 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)\C=C\C

InChI

1S/C5H8O2/c1-3-4-5(6)7-2/h3-4H,1-2H3/b4-3+

InChI key

MCVVUJPXSBQTRZ-ONEGZZNKSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

30.2 °F - closed cup

flash_point_c

-1 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library