Skip to Content
Merck

Skip To

94445

Mannose Disks

suitable for microbiology, Sterile filter paper discs impregnated with mannose

Synonym(s):

Carbohydrate discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 x 25 discs
Please contact Customer Service for Availability
2 740,00 CZK

About This Item

NACRES:
NA.85
UNSPSC Code:
41171621
Sterility:
sterile
Packaging:
pkg of 10 × 25 discs

2 740,00 CZK


Please contact Customer Service for Availability

​
Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

storage temp.

2-8°C

suitability

Citrobacter spp., Enterobacter spp., Escherichia coli, Klebsiella spp., Salmonella spp., Serratia spp., Shigella spp., bacteria, coliforms

General description

The mannose fermentation test is based on the fermentation ability of mannose which results in the production of acid as end product, the pH of the liquid basal medium will drop and an indicator change the color to red. Some organisms produce additional CO2 gas which can be made visible with a Durham’s tube. Mannose can be used to differentiate Enterobacteriaceae and other organisms.

Application

Mannose Discs are recommended to detect mannose fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
944389399993196
description

suitable for microbiology, Sterile filter paper discs impregnated with mannose

description

suitable for microbiology, Sterile filter paper discs impregnated with mannitol

description

suitable for microbiology, Sterile filter paper discs impregnated with rhamnose

description

suitable for microbiology, Sterile filter paper discs impregnated with melibiose

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

packaging

pkg of 10 × 25 discs

form

disc

form

disc

form

disc

form

disc

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

application(s)

microbiology, clinical testing
environmental
food and beverages
pharmaceutical

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

application(s)

clinical testing
environmental
food and beverages
pharmaceutical, microbiology

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

product line

BioChemika

product line

BioChemika

product line

BioChemika

product line

BioChemika


Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters