Skip to Content
Merck
All Photos(1)

Documents

94265

Supelco

Uranyl ionophore I

Selectophore

Synonym(s):

ETH 295, N,N′-Diheptyl-N,N′,6,6-tetramethyl-4,8-dioxaundecanediamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H54N2O4
CAS Number:
Molecular Weight:
470.73
Beilstein:
2305577
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

SMILES string

CCCCCCCN(C)C(=O)CCOCC(C)(C)COCCC(=O)N(C)CCCCCCC

InChI

1S/C27H54N2O4/c1-7-9-11-13-15-19-28(5)25(30)17-21-32-23-27(3,4)24-33-22-18-26(31)29(6)20-16-14-12-10-8-2/h7-24H2,1-6H3

InChI key

UZKSGXIBTHOXEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore employed in electrodes for the detn. of uranyl

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Senkyr et al.
Analytical Chemistry, 51, 786-786 (1979)
P.A. Bertrand et al.
Analytical Chemistry, 55, 364-364 (1983)
E. Malinowska
Analyst, 115, 1085-1085 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service