Skip to Content
Merck
All Photos(1)

Documents

92642

Supelco

Lewatit® Sybron Ionac® SR-7

15-50 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

form

beads

Quality Level

technique(s)

LPLC: suitable

loss

~60% loss on drying

matrix active group

polymer

particle size

15-50 μm

separation technique

reversed phase

Features and Benefits

very high selectivity for nitrate ions; selective removal of nitrates

Legal Information

Ionac is a registered trademark of Sybron Chemical Holdings Inc.
Lewatit is a registered trademark of Lanxess Deutschland GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service