91403
Isovaleryl-L-carnitine
analytical standard
Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-(3-methyl-1-oxobutoxy)-1-propanaminium inner salt, L-Carnitine isovaleryl ester, iC5-Carnitine
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥94.0% (HPLC)
optical activity
[α]/D -23.0±2.0°, c = 0.1 in H2O
shelf life
limited shelf life, expiry date on the label
impurities
≤12% water (calculated from elemental analysis)
application(s)
clinical testing
format
neat
storage temp.
2-8°C
SMILES string
C[N+](C)(C)C[C@H](OC(CC(C)C)=O)CC([O-])=O
InChI
1S/C12H23NO4/c1-9(2)6-12(16)17-10(7-11(14)15)8-13(3,4)5/h9-10H,6-8H2,1-5H3/t10-/m1/s1
InChI key
IGQBPDJNUXPEMT-SNVBAGLBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service