Skip to Content
Merck
All Photos(3)

Key Documents

902284

Sigma-Aldrich

TyrannoPhos

Synonym(s):

Cyclopentylbis(3,5-di-tert-butylphenyl)phosphine, Phosphine ligand for nickel catalysis

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CZK 5,292.00
1 G
CZK 18,500.00

CZK 5,292.00

List PriceCZK 7,560.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
CZK 5,292.00
1 G
CZK 18,500.00

About This Item

Empirical Formula (Hill Notation):
C33H51P
CAS Number:
Molecular Weight:
478.73
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

CZK 5,292.00

List PriceCZK 7,560.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

functional group

phosphine

SMILES string

CC(C1=CC(P(C2=CC(C(C)(C)C)=CC(C(C)(C)C)=C2)C3CCCC3)=CC(C(C)(C)C)=C1)(C)C

InChI

1S/C33H51P/c1-30(2,3)23-17-24(31(4,5)6)20-28(19-23)34(27-15-13-14-16-27)29-21-25(32(7,8)9)18-26(22-29)33(10,11)12/h17-22,27H,13-16H2,1-12H3

InChI key

IWPJBOKLLTVJMY-UHFFFAOYSA-N

Application

This new aryl alkylphosphine ligand developed in Abby′s Doyle′s lab has been tailored specifically for nickel catalysis as demonstrated in the Suzuki coupling of acetals with boronic acids to generate benzylic ethers, reactions that pose challenges in Ni catalysis when using phosphine ligands developed for Pd-catalysed couplings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Wu et al.
Nature chemistry, 9(8), 779-784 (2017-07-30)
The field of Ni-catalysed cross-coupling has seen rapid recent growth because of the low cost of Ni, its earth abundance, and its ability to promote unique cross-coupling reactions. Whereas advances in the related field of Pd-catalysed cross-coupling have been driven

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service