Skip to Content
Merck
All Photos(1)

Key Documents

902225

Ir[dFMeppy]2-(4,4′-dCF3bpy)PF6

≥95%

Synonym(s):

Photocatalyst

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 2,770.00

CZK 2,770.00

List PriceCZK 5,540.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CZK 2,770.00

About This Item

Empirical Formula (Hill Notation):
C36H22F16IrN4P
CAS Number:
Molecular Weight:
1037.75
MDL number:
UNSPSC Code:
41141700
NACRES:
NA.22

CZK 2,770.00

List PriceCZK 5,540.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

core: iridium
reaction type: Photocatalysis
reagent type: catalyst

mp

>300 °C

SMILES string

FC1=CC(F)=C2C3=CC=C(C=[N]3[Ir+3]45([C-]2=C1)([C-]6=CC(F)=CC(F)=C6C7=CC=C(C=[N]74)C)[N]8=CC=C(C=C8C=9C=C(C=C[N]95)C(F)(F)F)C(F)(F)F)C.[F-][P+5]([F-])([F-])([F-])([F-])[F-]

InChI

InChI=1S/C12H6F6N2.2C12H8F2N.F6P.Ir/c13-11(14,15)7-1-3-19-9(5-7)10-6-8(2-4-20-10)12(16,17)18;2*1-8-2-5-12(15-7-8)10-4-3-9(13)6-11(10)14;1-7(2,3,4,5)6;/h1-6H;2*2-3,5-7H,1H3;;/q;3*-1;+3

InChI key

RYKRFKQFYGHADE-UHFFFAOYSA-N

Application

Under a dual catalytic copper/photoredox manifold, the Mes-Umemoto reagent has been demonstrated to be an excellent photocatalyst for the trifluoromethylation of alkyl halides and aryl halides to yield alkyl-CF3 and aryl-CF3 in high yields. In both cases, these reactions exhibit wide substrate scope with good functional group tolerance. More specifically, a variety of 5-membered and 6-membered heteroaryl halides can be readily converted to the corresponding trifluoromethylheteroarenes under mild conditions. For trifluoromethylation reactions, use with dMesSCF3 (901466) and supersilanol (902489).

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chip Le et al.
Science (New York, N.Y.), 360(6392), 1010-1014 (2018-06-02)
Transition metal-catalyzed arene functionalization has been widely used for molecular synthesis over the past century. In this arena, copper catalysis has long been considered a privileged platform due to the propensity of high-valent copper to undergo reductive elimination with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service