Skip to Content
Merck
All Photos(1)

Key Documents

900642

Sigma-Aldrich

Ethyl alcohol, Pure

≥99.5%, ZerO2®, 200 proof, anhydrous

Synonym(s):

Degassed and low oxygen ethanol, Ethanol ZerO2®, Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
grade:
ZerO2®
bp:
78 °C (lit.)

Product Name

Ethyl alcohol, Pure, 200 proof, anhydrous, ZerO2®, ≥99.5%

grade

ZerO2®

Quality Level

vapor density

1.59 (vs air)

Assay

≥99.5%

form

liquid

quality

anhydrous

expl. lim.

3.1-27.7 % (lit.)

concentration

200 proof

dilution

(for analytical testing)

impurities

≤0.005% water
water

bp

78 °C (lit.)

mp

-114 °C (lit.)

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihua Chen et al.
Nature communications, 10(1), 1112-1112 (2019-03-09)
Further minimizing the defect state density in the semiconducting absorber is vital to boost the power conversion efficiency of solar cells approaching Shockley-Queisser limit. However, it lacks a general strategy to control the precursor chemistry for defects density reduction in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service