Skip to Content
Merck

87734

Tetramethylammonium hydroxide solution

purum, ~10% in methanol (T)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)4N(OH)
CAS Number:
Molecular Weight:
91.15
PubChem Substance ID:
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
3558708
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

concentration

~10% in methanol (T)

impurities

≤0.3% halides (as chloride)

refractive index

n20/D 1.351

anion traces

sulfate (SO42-): ≤1000 mg/kg

SMILES string

[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.H2O/c1-5(2,3)4;/h1-4H3;1H2/q+1;/p-1

InChI key

WGTYBPLFGIVFAS-UHFFFAOYSA-M



Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT RE 1 Dermal - STOT SE 1

target_organs

Eyes,Central nervous system, Liver,thymus

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.1 °F - closed cup

flash_point_c

10.6 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library