Skip to Content
Merck
All Photos(1)

Key Documents

87191

Supelco

Sodium 1-tetradecanesulfonate

suitable for ion pair chromatography, LiChropur, ≥99.0%

Synonym(s):

1-Tetradecanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13SO3Na
CAS Number:
Molecular Weight:
300.43
Beilstein:
3920141
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0%

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O (hot)

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCCCCCCCS([O-])(=O)=O

InChI

1S/C14H30O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-18(15,16)17;/h2-14H2,1H3,(H,15,16,17);/q;+1/p-1

InChI key

AYFACLKQYVTXNS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Wang et al.
Journal of colloid and interface science, 491, 64-71 (2016-12-26)
Thermoreversible ionogels formed by pseudogemini surfactants were prepared in protic ionic liquid, ethylammonium nitrate (EAN). Gemini-type supra-amphiphiles were formed by single-chain surfactants and bola-type molecules in a 2:1M ratio. The structures of aggregates including polymorphous lamellar structures and fibrous networks
Zhen Zhang et al.
Colloids and surfaces. B, Biointerfaces, 171, 647-655 (2018-08-15)
Understanding the assembly mechanisms of supramolecular architectures in nature is essential for the design and synthesis of novel biomaterials. In the work, self-assembly of gelatin-mono epoxy terminated polydimethylsiloxane polymer (PGG) controlled by electrostatic and hydrophobic interactions between gelatin and sodium
Manuela B T Sousa et al.
Food research international (Ottawa, Ont.), 114, 223-229 (2018-10-27)
In this work, the use of ion pair chromatography strategy in low pressure chromatographic flow systems is explored for the first time. The straightforward flow manifold encompassed a peristaltic pump, an injection valve and a 1 cm-length C18 monolithic column. The
Miriam Gochin et al.
Journal of medicinal chemistry, 52(14), 4338-4344 (2009-06-19)
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service