Skip to Content
Merck

85676

3-Stearoyl-sn-glycerol

≥99.0% (TLC)

Synonym(s):

(R)-Glycerol 1-stearate, L-α-Monostearin, L-α-Stearin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H42O4
CAS Number:
Molecular Weight:
358.56
UNSPSC Code:
12352211
EC Number:
238-880-5
Beilstein/REAXYS Number:
1728686
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (TLC)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@H](O)CO





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library