83071
1-(4-Pyridyl)pyridinium chloride hydrochloride
technical, ≥85% (AT)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
technical
Assay
≥85% (AT)
mp
166-170 °C (dec.) (lit.)
SMILES string
[Cl-].Cl[H].c1cc[n+](cc1)-c2ccncc2
InChI
1S/C10H9N2.2ClH/c1-2-8-12(9-3-1)10-4-6-11-7-5-10;;/h1-9H;2*1H/q+1;;/p-1
InChI key
QZOFFMRDIRXGKJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service