Skip to Content
Merck
All Photos(1)

Key Documents

82475

Sigma-Aldrich

Prostaglandin E2

≥99.0% (TLC)

Synonym(s):

(5Z,11α,13E,15S)-11,15-Dihydroxy-9-oxoprosta-5,13-dienoic acid, Dinoprostone, PGE2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Beilstein:
4709356
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99.0% (TLC)

solubility

acetone: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1

InChI key

XEYBRNLFEZDVAW-ARSRFYASSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.The effect of PGE2 on the immune system is mixed. It inhibits T cell activation in vitro, suggesting it is an immunosuppressant. However, in vivo, it appears to effect expansion of the Th17 subset and differentiation of the Th1 subset of T helper cells, marking it as an immunoactivator.

Other Notes

Differential effect of PGE2 on transforming growth factor-β, and insulin-induced collagen formation in lung fibroblasts

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fine et al.
The Journal of biological chemistry, 264(29), 16988-16991 (1989-10-15)
We examined the effect of prostaglandin (PG) E2 on transforming growth factor-beta (TGF-beta) and insulin-stimulated collagen formation in lung fibroblast cultures. TGF-beta increased type I collagen production 2-3-fold as determined by the densitometric analysis of autoradiograms from polyacrylamide gels and
Alexandra Medeiros et al.
Mediators of inflammation, 2012, 327568-327568 (2012-10-02)
The local and systemic production of prostaglandin E(2) (PGE(2)) and its actions in phagocytes lead to immunosuppressive conditions. PGE(2) is produced at high levels during inflammation, and its suppressive effects are caused by the ligation of the E prostanoid receptors
Sofia Eberstål et al.
International journal of cancer, 134(11), 2748-2753 (2013-11-19)
Immunotherapy has shown effectiveness against experimental malignant brain tumors, but the clinical results have been less convincing most likely due to immunosuppression. Prostaglandin E2 (PGE2 ) is the key immunosuppressive product of cyclooxygenase-2 (COX-2) and increased levels of PGE2 and
Yumeng Mao et al.
Cancer research, 73(13), 3877-3887 (2013-05-02)
Tumors can suppress the host immune system by employing a variety of cellular immune modulators, such as regulatory T cells, tumor-associated macrophages, and myeloid-derived suppressor cells (MDSC). In the peripheral blood of patients with advanced stage melanoma, there is an
Takayuki Nakagawa
Hearing research, 276(1-2), 27-33 (2011-02-08)
Prostaglandins are one of the major groups of chemical mediators in the mammalian body. Among prostaglandins, prostaglandin E2 (PGE2) is the most abundant prostanoid in humans and involved in regulating many different fundamental biological functions. PGE2 signaling is mediated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service