Skip to Content
Merck
All Photos(1)

Key Documents

80100

Supelco

Dibutyl phthalate

Selectophore, for ion-selective electrodes

Synonym(s):

n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
1914064
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

vapor density

9.6 (vs air)

vapor pressure

1 mmHg ( 147 °C)

product line

Selectophore

Assay

99.50%

form

liquid

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

color

clear

refractive index

n20/D 1.492 (lit.)
n20/D 1.493

bp

340 °C (lit.)

mp

−35 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl phthalate (DBP) belongs to phthalate group, and is a dialkyl or aryl/alkyl diesters of phthalic acid. It is mostly used as additive in cosmetics and skin care products. Generally, phthalate esters are hydrophobic compounds which displays strong potential for adsorption.
Visit our Sensor Applications portal to learn more.

Application

DBP may be used as plasticisers in studying the transcriptional activity of human sodium/iodide symporter.
Plasticizer

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systemic uptake of diethyl phthalate, dibutyl phthalate, and butyl paraben following whole-body topical application and reproductive and thyroid hormone levels in humans.
Janjua NR
Environmental Science & Technology, 41(15), 5564-5570 (2007)
Caterina S Wondergem et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(7), 625-632 (2020-01-26)
Synthesis methods to prepare lower transition metal catalysts and specifically Ni for Shell-Isolated Nanoparticle-Enhanced Raman Spectroscopy (SHINERS) are explored. Impregnation, colloidal deposition, and spark ablation have been investigated as suitable synthesis routes to prepare SHINERS-active Ni/Au@SiO2 catalyst/Shell-Isolated Nanoparticles (SHINs). Ni
Biodegradation of bis (2-ethylhexyl) phthalate in a soil slurry-sequencing batch reactor.
Juneson, Christopher, Owen P. Ward, and Ajay Singh.
Process. Biochem., 37.3, 305-313 (2001)
The promoter of the human sodium/iodide symporter responds to certain phthalate plasticisers.
Breous E, Wenzel A, Loos U.
Molecular and Cellular Endocrinology, 244(1-2), 75-78 (2005)
Human biological monitoring of diisononyl phthalate and diisodecyl phthalate: a review.
Saravanabhavan G, Murray J.
Journal of Environmental and Public Health, 1-11 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service