Skip to Content
Merck
All Photos(2)

Documents

79544

Supelco

Galactinol dihydrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
8665287
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

analyte chemical class(es)

oligosaccharides

technique(s)

GC/MS: suitable
HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-2-3(14)4(15)10(21)12(22-2)23-11-8(19)6(17)5(16)7(18)9(11)20;;/h2-21H,1H2;2*1H2/t2-,3+,4+,5-,6-,7+,8-,9-,10-,11-,12-;;/m1../s1

InChI key

HGCURVXTXVAIIR-ITYUJIADSA-N

General description

Galactinol dihydrate is a crystalline form of galactinol, which comprises of α-D-galactose and myo-inositol.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and Identification of O-?-D-Galactopyranosyl-myo-inositol and of myo-Inositol from Juice of the Sugar Beet (Beta Vulgaris)
Brown.JR and Serro.FR
Journal of the American Chemical Society, 75, 1040?1042-1040?1042 (1953)
Advances in Carbohydrate Chemistry, 9 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service