Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Please contact Customer Service for Availability | CZK 14,300.00 |
About This Item
Empirical Formula (Hill Notation):
C12H20N6O3S
CAS Number:
Molecular Weight:
328.39
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
23151816
MDL number:
grade
derivatization grade (HPLC)
Quality Segment
assay
≥95.0% (HPLC)
form
powder
quality
LiChropur™
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
storage temp.
2-8°C
SMILES string
CN(C)CCNS(=O)(=O)c1ccc(NCCN)c2nonc12
InChI
1S/C12H20N6O3S/c1-18(2)8-7-15-22(19,20)10-4-3-9(14-6-5-13)11-12(10)17-21-16-11/h3-4,14-15H,5-8,13H2,1-2H3
InChI key
QKTBJERNSIEPPM-UHFFFAOYSA-N
Application
4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole (DAABD-AE) is a new derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids (i.e., hexacosanoic, tetracosanoic, and docosanoic acids) in plasma.
4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole may be used as derivatization reagent for the trace analysis of carboxylic acids in saliva samples using liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS).[1]
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Legal Information
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life - | shelf life - |
| quality LiChropur™ | quality - | quality - | quality - |
| form powder | form liquid | form liquid | form liquid |
| grade derivatization grade (HPLC) | grade - | grade - | grade - |
| assay ≥95.0% (HPLC) | assay ≥98.0% (GC) | assay ≥99.0% (GC) | assay 99% |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole derivatization grade (HPLC), LiChropur™, ≥95.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/873/844/30d439dd-8ab1-47d6-a2fd-ef3fd65ac928/640/30d439dd-8ab1-47d6-a2fd-ef3fd65ac928.png)


