Skip to Content
Merck
All Photos(2)

Key Documents

78864

Supelco

1,3,6-Tri-O-galloyl-β-D-glucose

analytical standard

Synonym(s):

β-D-Glucopyranose 1,3,6-trigallate, β-D-Glucopyranose-1,3,6-tris(3,4,5-trihydroxybenzoate), 1,3,6-Tri-O-galloyl-β-D-glucopyranose, NP 005114, NSC 69861

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CZK 13,790.00

CZK 13,790.00

List PriceCZK 19,700.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CZK 13,790.00

About This Item

Empirical Formula (Hill Notation):
C27H24O18
CAS Number:
Molecular Weight:
636.47
Beilstein:
76215
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

CZK 13,790.00

List PriceCZK 19,700.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC(=O)c4cc(c(c(c4)O)O)O)O)OC(=O)c3cc(c(c(c3)O)O)O)O)OC(=O)c2cc(c(c(c2)O)O)O

InChI

1S/C27H24O18/c28-11-1-8(2-12(29)18(11)34)24(39)42-7-17-21(37)23(44-25(40)9-3-13(30)19(35)14(31)4-9)22(38)27(43-17)45-26(41)10-5-15(32)20(36)16(33)6-10/h1-6,17,21-23,27-38H,7H2/t17-,21-,22-,23+,27+/m1/s1

InChI key

RNKMOGIPOMVCHO-SJMVAQJGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service