Skip to Content
Merck

77635

Phenolphthalein disulfate tripotassium salt

≥99.0% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H13O11S2K3
CAS Number:
Molecular Weight:
646.77
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (TLC)

impurities

≤0.01% free phenolphthalein (UV)

SMILES string

[K+].[K+].[K+].OC(c1ccc(OS([O-])(=O)=O)cc1)(c2ccc(OS([O-])(=O)=O)cc2)c3ccccc3C([O-])=O

Other Notes

Substrate for sulphatase; Inhibitor of UDP-glucuronyl transferase from rat liver




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library