Skip to Content
Merck

77100

Diethyl pentylmalonate

purum, ≥98.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)4CH(COOC2H5)2
CAS Number:
Molecular Weight:
230.30
UNSPSC Code:
12352103
EC Number:
227-997-7
Beilstein/REAXYS Number:
609224
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.426

bp

254-255 °C

SMILES string

CCCCCC(C(=O)OCC)C(=O)OCC





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library