Skip to Content
Merck

Skip To

76952

Sodium 1-pentanesulfonate monohydrate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

1-Pentanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2.5 g
Please contact Customer Service for Availability
CZK 3,130.00
CZK 2,347.50
10 g
Please contact Customer Service for Availability
CZK 4,230.00
50 g
Please contact Customer Service for Availability
CZK 17,800.00

About This Item

Linear Formula:
CH3(CH2)4SO3Na · H2O
CAS Number:
Molecular Weight:
192.21
UNSPSC Code:
41115711
NACRES:
NB.21
PubChem Substance ID:
EC Number:
245-208-4
Beilstein/REAXYS Number:
3725147
MDL number:

CZK 2,347.50

List PriceCZK 3,130.00Save 25%

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

assay

≥99.0% (T)

form

sheet

quality

LiChropur

mol wt

192.21 g/mol

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1, λ: 220 nm Amax: 0.06, λ: 230 nm Amax: 0.04, λ: 260 nm Amax: 0.02, λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test, corresponds to standard for filter test

SMILES string

O.[Na+].CCCCCS([O-])(=O)=O

InChI

1S/C5H12O3S.Na.H2O/c1-2-3-4-5-9(6,7)8;;/h2-5H2,1H3,(H,6,7,8);;1H2/q;+1;/p-1

InChI key

FPQYXAFKHLSWTI-UHFFFAOYSA-M

General description

Sodium 1-pentanesulfonate monohydrate is an Ion pair reagent. It is a anionic surfactant which has opposite influence on aniline polymerization thermal effect. It also has highest and lowest enthalpy of polymerization values.

Application

Sodium pentanesulfonate is useful as a surfactant and reversed-phase ion-paring agent.
Sodium 1-pentanesulfonate monohydrate may be used to investigate its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy for its structural characterization.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
518337695551832
technique(s)

ion pair chromatography: suitable

technique(s)

ion pair chromatography: suitable

technique(s)

-

technique(s)

ion pair chromatography: suitable

suitability

corresponds to standard for RP gradient test, corresponds to standard for filter test

suitability

-

suitability

-

suitability

corresponds to standard for RP gradient test, corresponds to standard for filter test

form

sheet

form

solid

form

flakes

form

solid

assay

≥99.0% (T)

assay

≥99.0% (T)

assay

≥98.0% (T)

assay

≥99.0% (T)

mol wt

192.21 g/mol

mol wt

220.26 g/mol

mol wt

-

mol wt

220.26 g/mol

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters