Skip to Content
Merck
All Photos(1)

Key Documents

75069

Supelco

2-(Octyloxy)benzonitrile

Selectophore, ≥99.0% (GC)

Synonym(s):

2-Cyanophenyl octyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO
CAS Number:
Molecular Weight:
231.33
Beilstein:
4865692
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Assay

≥99.0% (GC)

refractive index

n20/D 1.506

density

0.96 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCOc1ccccc1C#N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Non-quenching plasticizer for optode membranes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.B.Papkovsky et al.
Analytica Chimica Acta, 337, 201-201 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service